accessibility menu, dialog, popup

UserName

CAS RN 638-51-7

IUPAC Name: hexyl nitrite
Synonyms: hexyl nitrite
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: SGRWGISGVDVSJV-UHFFFAOYSA-N
SMILES: CCCCCCON=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results