missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Formate 98.0+%, TCI America™

Catalog No. H127125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H127125ML 25 mL
H1271500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H127125ML Supplier TCI America Supplier No. H127125ML

Chemical Identifiers

CAS 629-33-4
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00046146
InChI Key OUGPMNMLWKSBRI-UHFFFAOYSA-N
Synonym n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
PubChem CID 61177
IUPAC Name hexyl formate
SMILES CCCCCCOC=O

Specifications

Melting Point -63°C
Color Colorless
Boiling Point 156°C
Quantity 25 mL
UN Number 3272
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Formate
EINECSNumber (2)-3050
RTECSNumber LQ8595000
TSCA Yes