accessibility menu, dialog, popup

UserName

H3COO

CAS RN 629-33-4

IUPAC Name: hexyl formate
Synonyms: hexyl formate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: OUGPMNMLWKSBRI-UHFFFAOYSA-N
SMILES: CCCCCCOC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results