missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Ether 98.0+%, TCI America™

Supplier:  TCI America H013825ML

 View more versions of this product

Catalog No. H013825ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hexyl Ether
-43°C
226°C
MFCD00009525
dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane
CCCCCCOCCCCCC
186.34
186.34
Liquid
112-58-3
Yellow
C12H26O
25 mL
BPIUIOXAFBGMNB-UHFFFAOYSA-N
1-(hexyloxy)hexane
8198
≥98.0% (GC)

Chemical Identifiers

112-58-3
186.34
BPIUIOXAFBGMNB-UHFFFAOYSA-N
8198
CCCCCCOCCCCCC
C12H26O
MFCD00009525
dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane
1-(hexyloxy)hexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KN8600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions