accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 112-58-3

IUPAC Name: 1-(hexyloxy)hexane
Synonyms: di-N-hexyl ether
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: BPIUIOXAFBGMNB-UHFFFAOYSA-N
SMILES: CCCCCCOCCCCCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)