missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Ether 98.0+%, TCI America™

Catalog No. H0138500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0138500ML 500 mL
H013825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0138500ML Supplier TCI America Supplier No. H0138500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-58-3
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00009525
InChI Key BPIUIOXAFBGMNB-UHFFFAOYSA-N
Synonym dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane
PubChem CID 8198
IUPAC Name 1-(hexyloxy)hexane
SMILES CCCCCCOCCCCCC

Specifications

Melting Point -43°C
Color Yellow
Boiling Point 226°C
Quantity 500 mL
Formula Weight 186.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Ether
RTECSNumber KN8600000
TSCA Yes