missing translation for 'onlineSavingsMsg'
Learn More

Hexyl alcohol, 99%, anhydrous, AcroSeal™

CAS: 111-27-3 | C6H14O | 102.18 g/mol

Supplier:  Thermo Scientific Chemicals 433860010

 View more versions of this product

Catalog No. AC433860010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexyl alcohol
-52°C
0.8136g/mL
59°C
98.5% min. (GC)
C6H14O
CH3(CH2)5OH
0.0005 % max.
15,4734
Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether
CCCCCCO
102.18
5.3 mPa.s (20°C)
102.18
Liquid
111-27-3
Colorless
156°C to 157°C
Authentic
AcroSeal™ Glass Bottle
1.4170 to 1.419
1 L
0.8136
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
hexan-1-ol
8103
CHEBI:87393
99%

Chemical Identifiers

111-27-3
102.18
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
CHEBI:87393
CCCCCCO
C6H14O
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
8103
hexan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Warning

EINECSNumber : 203-852-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only