missing translation for 'onlineSavingsMsg'
Learn More

Hexyl alcohol, 99%, anhydrous, AcroSeal™

Catalog No. p-4229240
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433861000 100 mL AcroSeal™ Glass Bottle
AC433860010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433861000 Supplier Thermo Scientific Chemicals Supplier No. 433861000
Only null left

CAS: 111-27-3 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point -52°C
Color Colorless
Density 0.8136g/mL
Boiling Point 156°C to 157°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4170 to 1.419
Linear Formula CH3(CH2)5OH
Quantity 100 mL
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.8136
Merck Index 15,4734
Solubility Information Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether
Viscosity 5.3 mPa.s (20°C)
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously wit
EINECSNumber 203-852-3

RUO – Research Use Only