missing translation for 'onlineSavingsMsg'
Learn More

Hexyl acetate, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187882
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020595 1 mL
111019305 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020595 Supplier Merck Emd Millipore Supplier No. 255391ML
Only null left

Hexyl acetate is a short-chain ester and an aromatic compound with a green note flavor, widely used in food industries.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 142-92-7
Density 0.87 g/mL (at 25°C)
Flash Point 56°C
Physical Form Neat
Linear Formula CH3 COO(CH2)5 CH3
MDL Number MFCD00009524
Melting Point -80°C
Boiling Point 168°C to 170°C
Synonym Capryl acetate
Type Analytical Standard
Quantity 1 mL
Formula Weight 144.21
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less