missing translation for 'onlineSavingsMsg'
Learn More

Hexyl 7-(Diethylamino)coumarin-3-carboxylate 98.0+%, TCI America™

Catalog No. H11451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11451G 1 g
H11455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11451G Supplier TCI America Supplier No. H11451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 851963-03-6
Molecular Formula C20H27NO4
Molecular Weight (g/mol) 345.439
MDL Number MFCD06797113
InChI Key TYEFUKSNTLMNBV-UHFFFAOYSA-N
Synonym 7-(Diethylamino)coumarin-3-carboxylic Acid Hexyl Ester
PubChem CID 44629745
IUPAC Name hexyl 7-(diethylamino)-2-oxochromene-3-carboxylate
SMILES CCCCCCOC(=O)C1=CC2=C(C=C(C=C2)N(CC)CC)OC1=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 345.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Hexyl 7-(Diethylamino)coumarin-3-carboxylate