accessibility menu, dialog, popup

UserName

H3COONCH3CH3OO

CAS RN 851963-03-6

IUPAC Name: hexyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate
Synonyms: hexyl 7-(diethylamino)-2-oxochromene-3-carboxylate
Molecular Weight (g/mol): 345.44
Molecular Formula: C20H27NO4
InChi Key: TYEFUKSNTLMNBV-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC2=CC=C(C=C2OC1=O)N(CC)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results