missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HEXIM1 Monoclonal Antibody (K40046_16G4), Invitrogen™

Catalog No. MA555786
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555786 50 μL
MA556166 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555786 Supplier Invitrogen™ Supplier No. MA555786
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: WGQQQRQLGK KKHRRRPSKK KRHWKPYYKL TWEEKKKFDE KQSLRASRIR AEMFAKGQPV APYNTTQFLM DDHDQEEPDL KTGLYSKRAA AKSDDTSDDD FMEEGGEEDG GSDGMGGDGS EFLQRDFSET YERYHTESLQ NMSKQELIKE YLELEKCLSR MEDENNRLRL ESKRLGGDDA RVRELELELD RLRAENLQLL

The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to malignancy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5' and 3' UTRs, have been described.

Specifications

Antigen HEXIM1
Applications Western Blot
Classification Monoclonal
Clone K40046_16G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene HEXIM1
Gene Accession No. O94992
Gene Alias 7330426E13Rik; Cardiac lineage protein 1; CLP1; Clp-1; EDG1; Estrogen down-regulated gene 1 protein; FLJ13562; hexamethylene bisacetamide inducible 1; hexamethylene bis-acetamide inducible 1; hexamethylene bisacetamide-inducible protein; hexamethylene bis-acetamide-inducible protein 1; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; HEXIM1; HGNC:24953; HIS1; HMBA-inducible; MAQ1; menage a quatre 1; menage a quatre protein 1; Protein HEXIM1
Gene Symbols HEXIM1
Host Species Mouse
Immunogen Recombinant human HEXIM1 (immunogen range is 141-340).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less