missing translation for 'onlineSavingsMsg'
Learn More

Hexestrol 98.0+%, TCI America™

Catalog No. p-7126247
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04941G 1 g
D049410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04941G Supplier TCI America Supplier No. D04941G
Only null left

Chemical Identifiers

CAS 84-16-2
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00068996
InChI Key PBBGSZCBWVPOOL-UHFFFAOYNA-N
Synonym hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
PubChem CID 192197
IUPAC Name 4-[4-(4-hydroxyphenyl)hexan-3-yl]phenol
SMILES CCC(C(CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 1 g
Formula Weight 270.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexestrol
RTECSNumber SL0560850
TSCA No