missing translation for 'onlineSavingsMsg'
Learn More

Hexestrol, 98+%, Thermo Scientific Chemicals

Catalog No. AAJ6088903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6088903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6088903 Supplier Thermo Scientific Chemicals Supplier No. J6088903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nonsteroidal synthetic estrogen

Nonsteroidal synthetic estrogen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84-16-2
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00068996
InChI Key PBBGSZCBWVPOOL-UHFFFAOYNA-N
Synonym hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
PubChem CID 192197
SMILES CCC(C(CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 186°C
Color White
Quantity 1 g
Beilstein 3209460
Merck Index 14,4701
IUPAC Name 4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol
Formula Weight 270.37
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Hexestrol
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 201-518-1
RTECSNumber SL0560850
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only