missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL
Description
Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Yellow |
| Solubility | Miscible with water |
| Quantity | 500 mL |
| Grade | Specpure |
| Chemical Name or Material | Hexavalent Chromium standard solution, Cr+6 1000μg/mL |
Safety and Handling
| Hazard Category | H340-H350-H360 |
| Hazard Statement | GHS H Statement H334-H340-H350-H360-H317-H208 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 232-143-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only