missing translation for 'onlineSavingsMsg'
Learn More

Hexatetracontane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015318
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015318 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015318 Supplier Merck Emd Millipore Supplier No. 52913250MG
Only null left
Petroleum

Specifications

CAS 7098-24-0
Physical Form Neat
Linear Formula CH3(CH2)44 CH3
MDL Number MFCD00042946
Melting Point 86°C to 91°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 647.24
Percent Purity ≥98.0% (GC)
Grade Analytical Standard