missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaphenyldisiloxane 96.0+%, TCI America™
$167.76 - $769.86
Chemical Identifiers
| CAS | 1829-40-9 |
|---|---|
| Molecular Formula | C36H30OSi2 |
| Molecular Weight (g/mol) | 534.805 |
| MDL Number | MFCD00014068 |
| InChI Key | IVZTVZJLMIHPEY-UHFFFAOYSA-N |
| PubChem CID | 74587 |
| IUPAC Name | triphenyl(triphenylsilyloxy)silane |
| SMILES | C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O[Si](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6 |
Chemical Identifiers
| 1829-40-9 | |
| 534.805 | |
| IVZTVZJLMIHPEY-UHFFFAOYSA-N | |
| triphenyl(triphenylsilyloxy)silane |
| C36H30OSi2 | |
| MFCD00014068 | |
| 74587 | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O[Si](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6 |
Specifications
| 1829-40-9 | |
| White | |
| MFCD00014068 | |
| IVZTVZJLMIHPEY-UHFFFAOYSA-N | |
| triphenyl(triphenylsilyloxy)silane | |
| 74587 | |
| ≥96.0% (GC) | |
| Hexaphenyldisiloxane |
| 228°C | |
| C36H30OSi2 | |
| 5 g | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O[Si](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6 | |
| 534.805 | |
| 534.81 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes