accessibility menu, dialog, popup

UserName

CAS RN 1829-40-9

OSiSi

CAS RN 1829-40-9

IUPAC Name: triphenyl[(triphenylsilyl)oxy]silane
Synonyms: triphenyl[(triphenylsilyl)oxy]silane
Molecular Weight (g/mol): 534.81
Molecular Formula: C36H30OSi2
InChi Key: IVZTVZJLMIHPEY-UHFFFAOYSA-N
SMILES: O([Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results