missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaphenyldisiloxane 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 228°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 534.81 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Hexaphenyldisiloxane |
Safety and Handling
| TSCA | Yes |