missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaphenoxycyclotriphosphazene 98.0+%, TCI America™
Supplier: TCI America H135625G
Specifications
| Hexaphenoxycyclotriphosphazene | |
| 114°C | |
| 280°C | |
| MFCD00183774 | |
| RNFJDJUURJAICM-UHFFFAOYSA-N | |
| 2,2,4,4,6,6-hexaphenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene | |
| 136917 | |
| ≥98.0% (HPLC,N) |
| 1184-10-7 | |
| White | |
| C36H30N3O6P3 | |
| 25 g | |
| C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(OC3=CC=CC=C3)OC4=CC=CC=C4)(OC5=CC=CC=C5)OC6=CC=CC=C6)OC7=CC=CC=C7 | |
| 693.572 | |
| 693.57 | |
| Crystalline Powder |
Chemical Identifiers
| 1184-10-7 | |
| 693.572 | |
| RNFJDJUURJAICM-UHFFFAOYSA-N | |
| 2,2,4,4,6,6-hexaphenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
| C36H30N3O6P3 | |
| MFCD00183774 | |
| 136917 | |
| C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(OC3=CC=CC=C3)OC4=CC=CC=C4)(OC5=CC=CC=C5)OC6=CC=CC=C6)OC7=CC=CC=C7 |
Safety and Handling
TSCA : No