missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoyl Chloride 98.0+%, TCI America™
$50.05 - $236.80
Chemical Identifiers
| CAS | 142-61-0 |
|---|---|
| Molecular Formula | C6H11ClO |
| Molecular Weight (g/mol) | 134.603 |
| MDL Number | MFCD00000760 |
| InChI Key | YWGHUJQYGPDNKT-UHFFFAOYSA-N |
| Synonym | caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid |
| PubChem CID | 67340 |
| IUPAC Name | hexanoyl chloride |
| SMILES | CCCCCC(=O)Cl |
Chemical Identifiers
| 142-61-0 | |
| 134.603 | |
| YWGHUJQYGPDNKT-UHFFFAOYSA-N | |
| 67340 | |
| CCCCCC(=O)Cl |
| C6H11ClO | |
| MFCD00000760 | |
| caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid | |
| hexanoyl chloride |
Specifications
| 142-61-0 | |
| 153°C | |
| MFCD00000760 | |
| 2920 | |
| YWGHUJQYGPDNKT-UHFFFAOYSA-N | |
| hexanoyl chloride | |
| 67340 | |
| ≥98.0% (GC,T) | |
| Hexanoyl Chloride |
| Yellow | |
| C6H11ClO | |
| 25 g | |
| caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid | |
| CCCCCC(=O)Cl | |
| 134.603 | |
| 134.60 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes