accessibility menu, dialog, popup

UserName

H3CClO

CAS RN 142-61-0

IUPAC Name: hexanoyl chloride
Synonyms: hexanoyl chloride
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: YWGHUJQYGPDNKT-UHFFFAOYSA-N
SMILES: CCCCCC(Cl)=O
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (5)