missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl Chloride 98.0+%, TCI America™

Catalog No. H011725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H011725G 25 g
H0117100G 100 g
H0117500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H011725G Supplier TCI America Supplier No. H011725G

Chemical Identifiers

CAS 142-61-0
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000760
InChI Key YWGHUJQYGPDNKT-UHFFFAOYSA-N
Synonym caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
PubChem CID 67340
IUPAC Name hexanoyl chloride
SMILES CCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 2920
Formula Weight 134.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexanoyl Chloride
EINECSNumber (2)-0632
TSCA Yes