missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl chloride, 97%

Catalog No. AC169041000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169041000 100 g Glass bottle
AC169045000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169041000 Supplier Thermo Scientific Chemicals Supplier No. 169041000

CAS: 142-61-0 | C6H11ClO | 134.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142-61-0
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.61
MDL Number MFCD00000760
InChI Key YWGHUJQYGPDNKT-UHFFFAOYSA-N
Synonym caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
PubChem CID 67340
IUPAC Name hexanoyl chloride
SMILES CCCCCC(=O)Cl

Specifications

Melting Point -87.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 151.0°C to 153.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4253 to 1.4273
Linear Formula CH3(CH2)4COCl
Quantity 100 g
Beilstein 02, 324
Specific Gravity 0.95
Merck Index 15, 1765
Solubility Information Solubility in water: hydrolysis. Other solubilities: decomposes in alcohol, soluble in ether, chloroform, ethylether and, acetone
Formula Weight 134.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Hexanoyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-549-1
TSCA TSCA

RUO – Research Use Only