missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic acid, For synthesis, MilliporeSigma™

Catalog No. M8001989026
Change view
Click to view available options
Quantity:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M8001989026 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8001989026 Supplier Technidata Supplier No. 8.00198.9026

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key FUZZWVXGSFPDMH-UHFFFAOYSA-N
Synonym caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid
PubChem CID 8892
ChEBI CHEBI:30776
IUPAC Name hexanoic acid
SMILES CCCCCC(=O)O

Specifications

Melting Point -3°C
Density 0.93g/3 (20°C)
pH 4 (1g/L, H2O, 20°C)
Boiling Point 206°C (1013 hPa)
Flash Point 110°C
Packaging Plastic Container
Quantity 25 L
Solubility Information 9.7g/L
Vapor Pressure 0.3 hPa (20°C)
Formula Weight 116.16
Percent Purity >98.0 % (a/a)
Show More Show Less
EINECSNumber 205-550-7
RTECSNumber MO5250000