missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, Reagent, ACS, 98.5%, Spectrum™ Chemical

H1010, 110-54-3

Supplier:  Spectrum Chemical Mfg Cor H10104LTGL

 View more versions of this product

Catalog No. 18603124


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Hexanes, Reagent, ACS , is a mixture of hexane isomers plus methylcyclopentane and is a colorless liquid. It is a used as non-polar solvent since it is low cost, largely unreactive, and relatively safe nature and can be found in specialized glue, as a degreasing agent and to extract cooking oil from seeds. It is used in the laboratory to remove or extract oil and grease contaminant from soil and water sample for analysis. Because it cannot easily be deprotonated it is often used for reactions that involve very strong bases. As an ACS grade Reagent, Hexanes manufactured hexanes is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Specifications

Specifications

Hexanes
100%
4 L
0.001%
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
98.5%
110-54-3
66°C to 70°C
Amber Glass Bottle
C6H14
UN1208
CCCCCC
86.18
Reagent

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
CCCCCC
C6H14
MFCD02179311
hexane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions