missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, Reagent, ACS, 98.5%, Spectrum™ Chemical

Catalog No. p-7157532
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603120 100 mL Amber Glass Bottle
18603121 500 mL Amber Glass Bottle
18603123 1 L Amber Glass Bottle
18603122 6 x 500 mL Amber Glass Bottle
18603124 4 L Amber Glass Bottle
18603125 6 x 1 L Amber Glass Bottle
18603126 4 x 4 L Amber Glass Bottle
18603127 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18603120 Supplier Spectrum Chemical Mfg Cor Supplier No. H1010100ML
Only null left

H1010, 110-54-3

Spectrum™ Chemical Hexanes, Reagent, ACS , is a mixture of hexane isomers plus methylcyclopentane and is a colorless liquid. It is a used as non-polar solvent since it is low cost, largely unreactive, and relatively safe nature and can be found in specialized glue, as a degreasing agent and to extract cooking oil from seeds. It is used in the laboratory to remove or extract oil and grease contaminant from soil and water sample for analysis. Because it cannot easily be deprotonated it is often used for reactions that involve very strong bases. As an ACS grade Reagent, Hexanes manufactured hexanes is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Max % 100%
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
UN Number UN1208
Boiling Point 66°C to 70°C
Percent Purity 98.5%
Grade Reagent
Chemical Name or Material Hexanes