missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, Practical, 85%, Spectrum™ Chemical

$256.53 - $6663.79

Chemical Identifiers

CAS 110-54-3
Molecular Weight (g/mol) 86.18
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610820
View Documents
Spectrum Chemical Mfg Cor
H1011500MLGL
500 mL Amber Glass Bottle
Each for $256.53
Only null left
 
18610819
View Documents
Spectrum Chemical Mfg Cor
H10114LTGL
4 L Amber Glass Bottle
Each for $580.47
Only null left
 
18610817
View Documents
Spectrum Chemical Mfg Cor
H101120LTMT
20 L Steel Can
Each for $1,648.65
Only null left
 
18610818
View Documents
Spectrum Chemical Mfg Cor
H1011200LTBL
200 L Drum
Each for $6,663.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-54-3
86.18

Specifications

110-54-3,96-37-7
66°C to 70°C
Amber Glass Bottle
0.85
Hexanes
40 to 60%,1 to 20%,40 to 60%
500 mL
86.18
Practical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.