missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, Practical, 85%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H1011200LTBL

 View more versions of this product

Catalog No. 18610818


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hexanes
40 to 60%,1 to 20%,40 to 60%
200 L
86.18
Practical
110-54-3,96-37-7
66°C to 70°C
Drum
0.85

Chemical Identifiers

110-54-3
86.18
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.