Learn More
Hexanes, mixed isomers, HPLC Grade, 99+%
CAS: 92112-69-1 | C6H14
Supplier: Thermo Scientific Chemicals 045652K7
Description
Hexanes, mixed isomers are used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexanes | |
| 92112-69-1,110-54-3,96-14-0,107-83-5,96-37-7,71-43-2 | |
| Colorless liquid | |
| 65°C to 70°C | |
| <-23°C (-9°F) | |
| Liquid | |
| >99% | |
| MFCD02179311 | |
| Miscible with ether,chloroform,alcohol,and acetone. Immiscible with water. | |
| HPLC |
| -95°C | |
| Colorless | |
| 0.67 g/cm3 | |
| 4 L | |
| 240°C (464°F) | |
| 1.3795 | |
| C6H14 | |
| 160 hPa (120mm Hg) at 20°C | |
| 86.18 | |
| n-Hexane: 65.0%; 3-Methylpentane: 25.0%; 2-Methylpentane: 5.0%; Methylcyclopentane: 4.999%; Benzene: 0.001% |
RUO – Research Use Only