missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, HPLC Grade, 99+%

Catalog No. p-4408266
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45652K2 1 L
AA45652M1 4 x 1 L
AA45652K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA45652K2 Supplier Thermo Scientific Chemicals Supplier No. 045652K2
Only null left

CAS: 92112-69-1 | C6H14

Hexanes, mixed isomers are used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Hexanes
Melting Point -95°C
CAS 92112-69-1, 110-54-3, 96-14-0, 107-83-5, 96-37-7, 71-43-2
Color Colorless
Appearance Colorless liquid
Density 0.67
Boiling Point 65°C to 70°C
Quantity 1 L
Flash Point <-23°C (-9°F)
Ignition Point 240°C (464°F)
Physical Form Liquid
Refractive Index 1.3795
Assay Percent Range >99%
Molecular Formula C6H14
Vapor Pressure 160 hPa (120mm Hg) at 20°C
MDL Number MFCD02179311
Solubility Information Miscible with ether,chloroform,alcohol,and acetone. Immiscible with water.
Formula Weight 86.18
Grade HPLC
Concentration or Composition (by Analyte or Components) n-Hexane: 65.0%; 3-Methylpentane: 25.0%; 2-Methylpentane: 5.0%; Methylcyclopentane: 4.999%; Benzene: 0.001%
Show More Show Less

RUO – Research Use Only