missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, Environmental Grade

Catalog No. AA42386K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42386K2 1 L
AA42386K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42386K2 Supplier Thermo Scientific Chemicals Supplier No. 042386K2

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. It is also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009520
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.67 g/mL
Quantity 1 L
Flash Point <−23°C (−9°F)
Physical Form Liquid
Refractive Index 1.3795
UN Number UN1208
Boiling Point 65°C to 70°C
Solubility Information Miscible with alcohol,chloroform,acetone and ether. Immiscible with water.
Formula Weight 86.18
Grade Environmental
Chemical Name or Material Hexanes, Mixed isomers
Show More Show Less
Hazard Category H225-H304-H315-H319-H335-H336-H361f-H372
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
EINECSNumber 295-570-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only