Learn More
Hexanes, mixed isomers, ACS, 98.5+%
CAS: 92112-69-1 | C6H14 | 86.18 g/mol
Supplier: Thermo Scientific Chemicals 033321K7
Description
Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexanes | |
| Mixed isomers | |
| Colorless | |
| 65°C to 70°C | |
| <-23°C (-9°F) | |
| 1.3795 | |
| MFCD00009520 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 | |
| ACS |
| -95°C | |
| 92112-69-1 | |
| 0.67 g/mL | |
| 4 L | |
| Liquid, Mixture Of 5 Isomers | |
| C6H14 | |
| UN1208 | |
| Miscible with alcohol,chloroform,acetone and ether. Immiscible with water. | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| ≥98.5% |
Chemical Identifiers
| 92112-69-1 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD00009520 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
EINECSNumber : 295-570-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only