missing translation for 'onlineSavingsMsg'
Learn More

Hexanes (Certified ACS), Fisher Chemical™

Catalog No. p-216058
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H2921 1 L Glass Bottle
H2924 4 L Glass Bottle
H29220 20 L Steel Pail
H292200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. H2921 Supplier Thermo Fisher Scientific Supplier No. H292 1
Only null left

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label.

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Quantity 1 L
Flash Point -6.7°C (+20°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Specific Gravity Less-than water
Assay Percent Range ≥98.5 %
UN Number 1208
Merck Index 15, 4732
Boiling Point 65.5°C
Solubility Information Negligible
Formula Weight 86.18
Percent Purity ≥98.5%
Grade Certified ACS
Chemical Name or Material Hexanes
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid