Learn More
Hexanes, 99%, ACS reagent, mixture of isomers
CAS: 92112-69-1 | C6H14
Supplier: Thermo Scientific Chemicals 423760025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexanes | |
| Mixture of isomers | |
| 0.6590g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.3748 to 1.3810 | |
| 99% | |
| 0.0003 meq/g max. | |
| CH3(CH2)4CH3 | |
| Vapor Pressure: 160mbar at 20°C | |
| 01, 142 | |
| Hex | |
| 86.18 | |
| ACS Reagent |
| -95.0°C | |
| 92112-69-1 | |
| 69.0°C | |
| −22°C | |
| Liquid | |
| 0.001% max. | |
| 0.659 | |
| C6H14 | |
| MFCD00009520 | |
| 0.31 mPa.s (20°C) | |
| 14, 4694 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
| 99% |
Chemical Identifiers
| 92112-69-1 | |
| C6H14 | |
| MFCD00009520 | |
| Hex |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.
Danger
EINECSNumber : 203-777-6
RTECSNumber : MN9275000
TSCA : TSCA
RUO – Research Use Only