missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, 99%, réactif ACS, mélange d’isomères
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | Hexanes |
| Point de fusion | -95,0 °C |
| Nom Remarque | Mélange d’isomères |
| Densité | 0.6590g/mL |
| Quantité | 500 mL |
| Point d’éclair | −22 °C |
| Emballage | EcoSafPak™ Bouteille en verre |
| Forme physique | Liquide |
| Indice de réfraction | 1,3748 à 1,3810 |
| Résidus après évaporation | 0,001% max. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. Suspected of damaging fertility. H |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 203-777-6 |
| RTECSNumber | MN9275000 |
| TSCA | TSCA |
RUO – Usage réservé à la recherche