missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, 99%, ACS reagent, mixture of isomers
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -95.0°C |
| Density | 0.6590g/mL |
| Quantity | 10 L |
| Flash Point | −22°C |
| Packaging | Plastic Drum |
| Physical Form | Liquid |
| Refractive Index | 1.3748 to 1.3810 |
| Residue after Evaporation | 0.001% max. |
| Specific Gravity | 0.659 |
| Assay Percent Range | 99% |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention. |
| EINECSNumber | 203-777-6 |
| RTECSNumber | MN9275000 |
| TSCA | TSCA |
RUO – Research Use Only