missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 99%, ACS reagent, mixture of isomers

Catalog No. AC423760010
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423760010 1 L EcoSafPak™ Glass Bottle
AC423765000 500 mL EcoSafPak™ Glass Bottle
AC423760025 2.5 L Glass Bottle
AC423760100 10 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423760010 Supplier Thermo Scientific Chemicals Supplier No. 423760010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
MDL Number MFCD00009520
Synonym Hex

Specifications

Melting Point -95.0°C
Density 0.6590g/mL
Quantity 1 L
Flash Point −22°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Residue after Evaporation 0.001% max.
Specific Gravity 0.659
Assay Percent Range 99%
Titratable Acid 0.0003 meq/g max.
Vapor Pressure Vapor Pressure: 160mbar at 20°C
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
Beilstein 01, 142
Merck Index 14, 4694
Boiling Point 69.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Percent Purity 99%
Grade ACS
Chemical Name or Material Hexanes, Mixture of isomers
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-777-6
RTECSNumber MN9275000
TSCA TSCA

RUO – Research Use Only