missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 98+%, extra pure, mixture of isomers

CAS: 92112-69-1 | C6H14

$113.37 - $195.12

Specifications

Chemical Name or Material Hexanes
Melting Point -95.0°C
Name Note Mixture of Isomers
CAS 92112-69-1 , 110-54-3
Color Colorless to Yellow
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC176660010
View Documents
Thermo Scientific Chemicals
176660010
1 L Glass Bottle
Each for $113.37
Only null left
 
AC176660025
View Documents
Thermo Scientific Chemicals
176660025
2.5 L Glass Bottle
Each for $195.12
Case of 4 Each for $0.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexanes
Mixture of Isomers
Colorless to Yellow
69.0°C
Liquid or Solid
98+%
0.659
CH3(CH2)4CH3
0.31 mPa.s (20°C)
0.05% Max.
Hex
86.18
Extra Pure
-95.0°C
92112-69-1 , 110-54-3
0.6590g/mL
−22°C
1.3748 to 1.3810
0.001 % max.
C6H14
MFCD00009520
01, 142
14, 4694
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
≥98%
Videos
SDS
Documents

Documents

RUO – Research Use Only