missing translation for 'onlineSavingsMsg'
Learn More

Heptane, CHROMASOLV™, ≥99%, LC-MS, Solstice

Catalog No. 6001024
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001024 1 L
6001025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001024 Supplier Solstice Supplier No. 349991L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Density 0.684g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Linear Formula CH3(CH2)5CH3
Vapor Pressure 83 mmHg (37.7°C), 40 mmHg (20°C)
UN Number UN1206
Beilstein 1730763
Formula Weight 100.2g/mol
Percent Purity ≥99%
Grade LC-MS
Chemical Name or Material Heptane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
RTECSNumber MI7700000