missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylenetetramine, 99% (Methenamine), Reagents
Hexamethylenetetramine, 99% (Methenamine), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C22136007KG
Specifications
| Hexamethylenetetramine, 99% (Methenamine) | |
| Sublimes | |
| Colorless/White | |
| 280°C | |
| C6H12N4 | |
| 1.331 | |
| VKYKSIONXSXAKP-UHFFFAOYSA-N | |
| 1,3,5,7-tetraazatricyclo[3.3.1.1³,â·]decane | |
| Reagent |
| 100-97-0 | |
| 280°C | |
| Sublimes | |
| Poly Pail | |
| 7 kg | |
| Methenamine, Urotropine, Hexamine | |
| C1N2CN3CN1CN(C2)C3 | |
| 140.19 | |
| Solid |
Chemical Identifiers
| 100-97-0 | |
| 140.19 | |
| Methenamine, Urotropine, Hexamine | |
| C1N2CN3CN1CN(C2)C3 |
| C6H12N4 | |
| VKYKSIONXSXAKP-UHFFFAOYSA-N | |
| 1,3,5,7-tetraazatricyclo[3.3.1.1³,â·]decane |