missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, 99% (Methenamine), Reagents

Catalog No. p-200020942
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
7 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337221 100 g
01337220 1 kg
01337219 2.5 kg
01337218 7 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337221 Supplier Labchem Inc Supplier No. C2213600100C2
Only null left

Hexamethylenetetramine, 99% (Methenamine), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym Methenamine, Urotropine, Hexamine
IUPAC Name 1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Boiling Range Sublimes
Melting Point 280°C
Color Colorless/White
Boiling Point Sublimes
Melting Range 280°C
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.331
Grade Reagent
Physical Form Solid
Chemical Name or Material Hexamethylenetetramine, 99% (Methenamine)
Show More Show Less