missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyleneammonium Hexamethylenedithiocarbamate 97.0+%, TCI America™

Catalog No. p-7130578
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05195G 5 g
H051925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05195G Supplier TCI America Supplier No. H05195G
Only null left

Chemical Identifiers

CAS 2608-11-9
Molecular Formula C13H26N2S2
Molecular Weight (g/mol) 274.49
MDL Number MFCD00036327
InChI Key CIPHOUPBLAOEKY-UHFFFAOYSA-N
Synonym Hexamethylenedithiocarbamic Acid Hexamethyleneammonium Salt
PubChem CID 164998
IUPAC Name azepane;azepane-1-carbodithioic acid
SMILES C1CCCNCC1.C1CCCN(CC1)C(=S)S

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Formula Weight 274.49
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethyleneammonium Hexamethylenedithiocarbamate