accessibility menu, dialog, popup

UserName

HSSNHN

CAS RN 2608-11-9

IUPAC Name: azepane; azepane-1-carbodithioic acid
Synonyms: azepane-1-carbodithioic acid; hexamethyleneimine
Molecular Weight (g/mol): 274.49
Molecular Formula: C13H26N2S2
InChi Key: CIPHOUPBLAOEKY-UHFFFAOYSA-N
SMILES: C1CCCNCC1.SC(=S)N1CCCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results