Learn More
Hexamethylbenzene, 99+%
CAS: 87-85-4 | C12H18 | 162.276 g/mol
Supplier: Thermo Scientific Chemicals A1029406
Description
Hexamethylbenzene is used as pharmaceutical intermediates, in organic synthesis of compounds, as a solvent for He-NMR spectroscopy. Used as a ligand in organometallic chemistry. Reaction with dimethyldioxirane, gives the major product, an unusual oxepane triepoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexamethylbenzene is used as pharmaceutical intermediates, in organic synthesis of compounds, as a solvent for He-NMR spectroscopy. Used as a ligand in organometallic chemistry. Reaction with dimethyldioxirane, gives the major product, an unusual oxepane triepoxide.
Solubility
Insoluble in water
Notes
Incompatible with nitromethane, strong oxidizing agents.
Specifications
| Hexamethylbenzene | |
| 163°C to 167°C | |
| 265°C | |
| MFCD00008523 | |
| 1905834 | |
| Insoluble in water | |
| CC1=C(C(=C(C(=C1C)C)C)C)C | |
| 162.276 | |
| CHEBI:39001 | |
| ≥99% |
| 87-85-4 | |
| 1.063 | |
| C12H18 | |
| 5 g | |
| hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene; | |
| YUWFEBAXEOLKSG-UHFFFAOYSA-N | |
| 1,2,3,4,5,6-hexamethylbenzene | |
| 6908 | |
| 162.28 |
Chemical Identifiers
| 87-85-4 | |
| 162.276 | |
| YUWFEBAXEOLKSG-UHFFFAOYSA-N | |
| 6908 | |
| 1,2,3,4,5,6-hexamethylbenzene |
| C12H18 | |
| MFCD00008523 | |
| hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene; | |
| CHEBI:39001 | |
| CC1=C(C(=C(C(=C1C)C)C)C)C |
Safety and Handling
EINECSNumber : 201-777-0
RTECSNumber : DA3200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only