missing translation for 'onlineSavingsMsg'
Learn More

γ-Hexalactone, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015261
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015261 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015261 Supplier Merck Emd Millipore Supplier No. 685541ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 695-06-7
Density 1.023 g/mL (at 25°C)
Flash Point 98°C - closed cup
Linear Formula C6H10O2
MDL Number MFCD00005401
Boiling Point 219°C
Synonym gamma-Caprolactone; gamma-Ethyl-gamma-butyrolactone
Type Analytical Standard
Quantity 1 mL
Formula Weight 114.14
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less