missing translation for 'onlineSavingsMsg'
Learn More

Hexakis(bromomethyl)benzene 98.0+%, TCI America™

Catalog No. H09011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09011G 1 g
H09015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09011G Supplier TCI America Supplier No. H09011G

Chemical Identifiers

CAS 3095-73-6
Molecular Formula C12H12Br6
Molecular Weight (g/mol) 635.652
MDL Number MFCD00182539
InChI Key XJOUCILNLRXRTF-UHFFFAOYSA-N
Synonym hexakis bromomethyl benzene, hexa bromomethyl benzene, 1,2,3,4,5,6-hexakis bromomethyl benzene, benzene, hexakis bromomethyl, acmc-1aijn, 10.14272/xjoucilnlrxrtf-uhfffaoysa-n, benzene,1,2,3,4,5,6-hexakis bromomethyl, doi:10.14272/xjoucilnlrxrtf-uhfffaoysa-n
PubChem CID 76540
IUPAC Name 1,2,3,4,5,6-hexakis(bromomethyl)benzene
SMILES C(C1=C(C(=C(C(=C1CBr)CBr)CBr)CBr)CBr)Br

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 635.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexakis(bromomethyl)benzene