missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexahydroxybenzene 98.0+%, TCI America™
Supplier: TCI America H06431G
Specifications
| Hexahydroxybenzene | |
| C6H6O6 | |
| 1 g | |
| OC1=C(O)C(O)=C(O)C(O)=C1O | |
| 174.11 | |
| 174.11 | |
| Crystalline Powder |
| 608-80-0 | |
| MFCD00013966 | |
| VWPUAXALDFFXJW-UHFFFAOYSA-N | |
| benzene-1,2,3,4,5,6-hexol | |
| 69102 | |
| ≥98.0% (T) |
Chemical Identifiers
| 608-80-0 | |
| 174.11 | |
| VWPUAXALDFFXJW-UHFFFAOYSA-N | |
| benzene-1,2,3,4,5,6-hexol |
| C6H6O6 | |
| MFCD00013966 | |
| 69102 | |
| OC1=C(O)C(O)=C(O)C(O)=C1O |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer