accessibility menu, dialog, popup

UserName

CAS RN 608-80-0

OHOHOHOHHOHO

CAS RN 608-80-0

IUPAC Name: benzene-1,2,3,4,5,6-hexol
Synonyms: benzenehexol
Molecular Weight (g/mol): 174.11
Molecular Formula: C6H6O6
InChi Key: VWPUAXALDFFXJW-UHFFFAOYSA-N
SMILES: OC1=C(O)C(O)=C(O)C(O)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results