missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorophosphoric acid, ca 60% w/w aq. soln.

CAS: 16940-81-1 | HPF6

$50.84 - $298.34

Chemical Identifiers

CAS 16940-81-1
Molecular Formula HPF6
MDL Number MFCD00011350
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL15728AC
View Documents
Thermo Scientific Chemicals
L15728AC
25 mL
Each for $50.84
Only null left
 
AAL15728AE
View Documents
Thermo Scientific Chemicals
L15728AE
100 mL
Each for $123.22
Only null left
 
AAL15728AP
View Documents
Thermo Scientific Chemicals
L15728AP
500 mL
Each for $298.34
Only null left
 
Description

Description

Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, glass, alkali metals, powdered metals, strong oxidizing agents, metals and reacts violently with water.  
Specifications

Chemical Identifiers

16940-81-1
HPF6
MFCD00011350

Specifications

16940-81-1
HPF6
25 mL
Immiscible with water.
≈60% w/w aq. soln.
1.65
MFCD00011350
UN1782
145.97
Hexafluorophosphoric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H310+H330-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HEXAFLUOROPHOSPHORIC ACID

EINECSNumber : 241-006-5

RTECSNumber : SY7115000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only